Assay Details
| Gene Symbol: ERF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112922.3 | NP_188666.1 | 1 | 1946 | 147 | |
| GenBank mRNA | AY037254.1 | - | 1 | 1857 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF7| Gene Symbol: ERF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112922.3 | NP_188666.1 | 1 | 1946 | 147 | |
| GenBank mRNA | AY037254.1 | - | 1 | 1857 | 147 | |
Set Membership: |
|