Assay Details
| Gene Symbol: AT3G20380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112929.3 | NP_188673.1 | 1 - 2 | 207 | 80 | |
| GenBank mRNA | AK176197.1 | - | 1 - 2 | 152 | 80 | |
| AK176301.1 | - | 1 - 2 | 152 | 80 | ||
| AK176307.1 | - | 1 - 2 | 152 | 80 | ||
More Information
Set Membership: |
|

AT3G20380
Genomic Map