Assay Details
| Gene Symbol: AT3G20390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338478.1 | NP_001327365.1 | 4 - 5 | 656 | 97 | |
| NM_112930.3 | NP_188674.1 | 4 - 5 | 559 | 97 | ||
| GenBank mRNA | AF375446.1 | - | 4 - 5 | 494 | 97 | |
| AK227774.1 | - | 4 - 5 | 527 | 97 | ||
| AK229768.1 | - | 8 - 9 | 1514 | 97 | ||
| AY060547.1 | - | 4 - 5 | 422 | 97 | ||
| AY086036.1 | - | 4 - 5 | 551 | 97 | ||
More Information
Set Membership: |
|

AT3G20390
Genomic Map