Assay Details
| Gene Symbol: EMB2729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161157.2 | NP_001154629.1 | 16 - 17 | 2427 | 91 | |
| NM_112935.2 | NP_188679.2 | 15 - 16 | 2323 | 91 | ||
| GenBank mRNA | AK118785.1 | - | 15 - 16 | 2323 | 91 | |
| GQ374452.1 | - | 16 - 17 | 2368 | 91 | ||
More Information
Set Membership: |
|

EMB2729
Genomic Map