Assay Details
| Gene Symbol: EMB2729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161157.2 | NP_001154629.1 | 19 - 20 | 2696 | 105 | |
| NM_112935.2 | NP_188679.2 | 18 - 19 | 2592 | 105 | ||
| GenBank mRNA | AK118785.1 | - | 18 - 19 | 2592 | 105 | |
| GQ374452.1 | - | 19 - 20 | 2637 | 105 | ||
More Information
Set Membership: |
|

EMB2729
Genomic Map