Assay Details
| Gene Symbol: SVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112944.3 | NP_188688.1 | 7 - 8 | 1512 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SVL3| Gene Symbol: SVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112944.3 | NP_188688.1 | 7 - 8 | 1512 | 73 | |
Set Membership: |
|