Assay Details
| Gene Symbol: CAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036714.5 | NP_001031791.1 | 4 - 5 | 1237 | 100 | |
| NM_001342324.1 | NP_001328611.1 | 3 - 4 | 1697 | 100 | ||
| NM_119675.4 | NP_195235.1 | 4 - 5 | 1455 | 100 | ||
| GenBank mRNA | AY074301.1 | - | 4 - 5 | 1238 | 100 | |
| AY113854.1 | - | 3 - 4 | 1166 | 100 | ||
More Information
Set Membership: |
|

CAT2
Genomic Map