Assay Details
| Gene Symbol: CAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036714.5 | NP_001031791.1 | 6 - 7 | 1399 | 80 | |
| NM_001342324.1 | NP_001328611.1 | 5 - 6 | 1859 | 80 | ||
| NM_119675.4 | NP_195235.1 | 6 - 7 | 1617 | 80 | ||
| GenBank mRNA | AY074301.1 | - | 6 - 7 | 1400 | 80 | |
| AY113854.1 | - | 5 - 6 | 1328 | 80 | ||
More Information
Set Membership: |
|

CAT2
Genomic Map