Assay Details
| Gene Symbol: PIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203991.1 | NP_001190920.1 | 2 - 3 | 703 | 69 | |
| NM_119676.4 | NP_195236.1 | 2 - 3 | 894 | 69 | ||
| GenBank mRNA | AB493718.1 | - | 2 - 3 | 583 | 69 | |
| AF412110.1 | - | 2 - 3 | 632 | 69 | ||
| AY062803.1 | - | 2 - 3 | 630 | 69 | ||
| AY081580.1 | - | 2 - 3 | 583 | 69 | ||
| AY088485.1 | - | 2 - 3 | 632 | 69 | ||
More Information
Set Membership: |
|

PIP3
Genomic Map