Assay Details
| Gene Symbol: BSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119689.4 | NP_567980.1 | 4 - 5 | 1053 | 126 | |
| GenBank mRNA | AF439824.1 | - | 4 - 5 | 786 | 126 | |
| AY142063.1 | - | 4 - 5 | 736 | 126 | ||
More Information
Set Membership: |
|

BSK1
Genomic Map