Assay Details
| Gene Symbol: DAZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119694.2 | NP_195254.1 | 1 | 805 | 81 | |
| GenBank mRNA | AB493719.1 | - | 1 | 805 | 81 | |
| DQ446896.1 | - | 1 | 805 | 81 | ||
More Information
Set Membership: |
|

DAZ2
Genomic Map