Assay Details
| Gene Symbol: DRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119708.3 | NP_195268.2 | 4 - 5 | 644 | 93 | |
| GenBank mRNA | BT022119.1 | - | 4 - 5 | 640 | 93 | |
| BT025661.1 | - | 4 - 5 | 636 | 93 | ||
More Information
Set Membership: |
|

DRL1
Genomic Map