Assay Details
| Gene Symbol: DRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119708.3 | NP_195268.2 | 5 - 6 | 838 | 78 | |
| GenBank mRNA | BT022119.1 | - | 5 - 6 | 834 | 78 | |
| BT025661.1 | - | 5 - 6 | 830 | 78 | ||
More Information
Set Membership: |
|

DRL1
Genomic Map