Assay Details
| Gene Symbol: CLC-E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203995.1 | NP_001190924.1 | 3 - 4 | 1385 | 93 | |
| NM_119709.1 | NP_567985.1 | 3 - 4 | 1385 | 93 | ||
More Information
Set Membership: |
|

CLC-E
Genomic Map