Assay Details
| Gene Symbol: AT4G35730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119739.4 | NP_195298.2 | 3 - 4 | 441 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G35730| Gene Symbol: AT4G35730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119739.4 | NP_195298.2 | 3 - 4 | 441 | 132 | |
Set Membership: |
|