Assay Details
| Gene Symbol: SEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085027.2 | NP_001078496.1 | 3 - 4 | 581 | 82 | |
| NM_001085028.1 | NP_001078497.1 | 3 - 4 | 557 | 82 | ||
| NM_001342370.1 | NP_001328859.1 | 2 - 3 | 647 | 82 | ||
| NM_119743.4 | NP_195302.1 | 3 - 4 | 620 | 82 | ||
| GenBank mRNA | AY078973.1 | - | 3 - 4 | 476 | 82 | |
| AY091671.1 | - | 3 - 4 | 425 | 82 | ||
| U26945.1 | - | 3 - 4 | 459 | 82 | ||
More Information
Set Membership: |
|

SEN1
Genomic Map