Assay Details
| Gene Symbol: STY17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119744.5 | NP_195303.2 | 12 - 13 | 1670 | 63 | |
| GenBank mRNA | AY093017.1 | - | 12 - 13 | 1644 | 63 | |
| AY128938.1 | - | 12 - 13 | 1455 | 63 | ||
More Information
Set Membership: |
|

STY17
Genomic Map