Assay Details
| Gene Symbol: STY17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119744.5 | NP_195303.2 | 3 - 4 | 743 | 78 | |
| GenBank mRNA | AY093017.1 | - | 3 - 4 | 717 | 78 | |
| AY128938.1 | - | 3 - 4 | 528 | 78 | ||
More Information
Set Membership: |
|

STY17
Genomic Map