Assay Details
| Gene Symbol: NRPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342374.1 | NP_001329346.1 | 8 - 9 | 1902 | 102 | |
| NM_119746.4 | NP_195305.2 | 8 - 9 | 1902 | 102 | ||
More Information
Set Membership: |
|

NRPB1
Genomic Map