Assay Details
| Gene Symbol: AT4G35820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119748.1 | NP_195307.1 | 4 - 5 | 637 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G35820| Gene Symbol: AT4G35820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119748.1 | NP_195307.1 | 4 - 5 | 637 | 96 | |
Set Membership: |
|