Assay Details
| Gene Symbol: LARP1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342378.1 | NP_001328621.1 | 2 - 3 | 1351 | 90 | |
| NM_119755.3 | NP_567991.1 | 2 - 3 | 1351 | 90 | ||
| GenBank mRNA | AF370207.1 | - | 2 - 3 | 1302 | 90 | |
| AY133869.1 | - | 2 - 3 | 1009 | 90 | ||
More Information
Set Membership: |
|

LARP1c
Genomic Map