Assay Details
| Gene Symbol: LARP1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119755.3 | NP_567991.1 | 4 - 5 | 1580 | 93 | |
| GenBank mRNA | AF370207.1 | - | 4 - 5 | 1531 | 93 | |
| AY133869.1 | - | 4 - 5 | 1238 | 93 | ||
More Information
Set Membership: |
|

LARP1c
Genomic Map