Assay Details
| Gene Symbol: LARP1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342378.1 | NP_001328621.1 | 4 - 5 | 2156 | 76 | |
| NM_119755.3 | NP_567991.1 | 5 - 6 | 1645 | 76 | ||
| GenBank mRNA | AF370207.1 | - | 5 - 6 | 1596 | 76 | |
| AY133869.1 | - | 5 - 6 | 1303 | 76 | ||
More Information
Set Membership: |
|

LARP1c
Genomic Map