Assay Details
| Gene Symbol: APX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342391.1 | NP_001328787.1 | 2 - 3 | 279 | 76 | |
| NM_119763.3 | NP_195321.1 | 2 - 3 | 299 | 76 | ||
| GenBank mRNA | AK119023.1 | - | 2 - 3 | 279 | 76 | |
| BT006053.1 | - | 2 - 3 | 234 | 76 | ||
More Information
Set Membership: |
|

APX5
Genomic Map