Assay Details
| Gene Symbol: FAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119790.3 | NP_195345.1 | 2 - 3 | 1097 | 74 | |
| GenBank mRNA | U38416.1 | - | 2 - 3 | 955 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAH1| Gene Symbol: FAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119790.3 | NP_195345.1 | 2 - 3 | 1097 | 74 | |
| GenBank mRNA | U38416.1 | - | 2 - 3 | 955 | 74 | |
Set Membership: |
|