Assay Details
| Gene Symbol: AT4G36550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119818.6 | NP_195373.6 | 5 - 6 | 982 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G36550| Gene Symbol: AT4G36550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119818.6 | NP_195373.6 | 5 - 6 | 982 | 74 | |
Set Membership: |
|