Assay Details
| Gene Symbol: HB40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342415.1 | NP_001329011.1 | 1 - 2 | 146 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HB40| Gene Symbol: HB40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342415.1 | NP_001329011.1 | 1 - 2 | 146 | 97 | |
Set Membership: |
|