Assay Details
| Gene Symbol: APP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342416.1 | NP_001328936.1 | 6 - 7 | 1134 | 87 | |
| NM_001342417.1 | NP_001328935.1 | 6 - 7 | 1134 | 87 | ||
| NM_119840.7 | NP_195394.4 | 6 - 7 | 1214 | 87 | ||
More Information
Set Membership: |
|

APP1
Genomic Map