Assay Details
| Gene Symbol: APP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342416.1 | NP_001328936.1 | 8 - 9 | 1264 | 88 | |
| NM_001342417.1 | NP_001328935.1 | 8 - 9 | 1264 | 88 | ||
| NM_119840.7 | NP_195394.4 | 8 - 9 | 1344 | 88 | ||
More Information
Set Membership: |
|

APP1
Genomic Map