Assay Details
| Gene Symbol: PLP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204013.1 | NP_001190942.2 | 3 - 4 | 601 | 70 | |
| NM_119869.2 | NP_195423.1 | 3 - 4 | 735 | 70 | ||
More Information
Set Membership: |
|

PLP5
Genomic Map