Assay Details
| Gene Symbol: PLP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204013.1 | NP_001190942.2 | 4 - 5 | 764 | 88 | |
| NM_119869.2 | NP_195423.1 | 4 - 5 | 898 | 88 | ||
More Information
Set Membership: |
|

PLP5
Genomic Map