Assay Details
| Gene Symbol: CYCB1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119913.3 | NP_195465.1 | 4 - 5 | 627 | 92 | |
| GenBank mRNA | BT029006.1 | - | 4 - 5 | 527 | 92 | |
More Information
Set Membership: |
|

CYCB1;1
Genomic Map