Assay Details
| Gene Symbol: BT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119924.3 | NP_568031.1 | 2 - 3 | 434 | 149 | |
| GenBank mRNA | AY316678.1 | - | 1 - 2 | 228 | 149 | |
| BT025247.1 | - | 1 - 2 | 228 | 149 | ||
More Information
Set Membership: |
|

BT5
Genomic Map