Assay Details
| Gene Symbol: AGL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342463.1 | NP_001328890.1 | 1 - 2 | 528 | 78 | |
| NM_001342464.1 | NP_001328888.1 | 1 - 2 | 180 | 78 | ||
| NM_119955.3 | NP_195507.1 | 1 - 2 | 528 | 78 | ||
| GenBank mRNA | AF336979.1 | - | 1 - 2 | 180 | 78 | |
| BT024846.1 | - | 1 - 2 | 180 | 78 | ||
More Information
Set Membership: |
|

AGL21
Genomic Map