Assay Details
| Gene Symbol: AGL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342462.1 | NP_001328889.1 | 3 - 4 | 1063 | 137 | |
| NM_119955.3 | NP_195507.1 | 4 - 5 | 770 | 137 | ||
| GenBank mRNA | AF336979.1 | - | 4 - 5 | 422 | 137 | |
| BT024846.1 | - | 4 - 5 | 422 | 137 | ||
More Information
Set Membership: |
|

AGL21
Genomic Map