Assay Details
| Gene Symbol: CAD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342466.1 | NP_001327960.1 | 2 - 3 | 348 | 131 | |
| NM_001342467.1 | NP_001327959.1 | 2 - 3 | 348 | 131 | ||
| NM_001342468.1 | NP_001327958.1 | 2 - 3 | 238 | 131 | ||
| NM_119958.4 | NP_195510.1 | 2 - 3 | 348 | 131 | ||
| GenBank mRNA | AY302075.1 | - | 2 - 3 | 214 | 131 | |
More Information
Set Membership: |
|

CAD6
Genomic Map