Assay Details
| Gene Symbol: ELI3-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119960.3 | NP_195512.1 | 2 - 3 | 525 | 97 | |
| GenBank mRNA | AF361859.1 | - | 2 - 3 | 270 | 97 | |
| AY129478.1 | - | 2 - 3 | 198 | 97 | ||
| AY302080.1 | - | 2 - 3 | 198 | 97 | ||
| X67815.1 | - | 243 | 97 | |||
More Information
Set Membership: |
|

ELI3-2
Genomic Map