Assay Details
| Gene Symbol: ELI3-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119960.3 | NP_195512.1 | 3 - 4 | 1194 | 85 | |
| GenBank mRNA | AF361859.1 | - | 3 - 4 | 939 | 85 | |
| AY129478.1 | - | 3 - 4 | 867 | 85 | ||
| AY302080.1 | - | 3 - 4 | 867 | 85 | ||
| X67815.1 | - | 912 | 85 | |||
More Information
Set Membership: |
|

ELI3-2
Genomic Map