Assay Details
| Gene Symbol: AT4G38030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119964.2 | NP_195516.2 | 8 - 9 | 1034 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G38030| Gene Symbol: AT4G38030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119964.2 | NP_195516.2 | 8 - 9 | 1034 | 76 | |
Set Membership: |
|