Assay Details
| Gene Symbol: HD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085042.1 | NP_001078511.1 | 2 - 3 | 717 | 125 | |
| NM_119974.4 | NP_195526.1 | 2 - 3 | 735 | 125 | ||
| GenBank mRNA | AK226389.1 | - | 2 - 3 | 709 | 125 | |
| AY093153.1 | - | 2 - 3 | 685 | 125 | ||
| BT008873.1 | - | 1 - 2 | 524 | 125 | ||
More Information
Set Membership: |
|

HD1
Genomic Map