Assay Details
| Gene Symbol: HD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085042.1 | NP_001078511.1 | 5 - 6 | 1459 | 90 | |
| NM_119974.4 | NP_195526.1 | 5 - 6 | 1477 | 90 | ||
| GenBank mRNA | AK226389.1 | - | 5 - 6 | 1451 | 90 | |
| AY093153.1 | - | 5 - 6 | 1427 | 90 | ||
| BT008873.1 | - | 4 - 5 | 1266 | 90 | ||
More Information
Set Membership: |
|

HD1
Genomic Map