Assay Details
| Gene Symbol: HD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119974.4 | NP_195526.1 | 7 - 8 | 1607 | 61 | |
| GenBank mRNA | AK226389.1 | - | 7 - 8 | 1581 | 61 | |
| AY093153.1 | - | 7 - 8 | 1557 | 61 | ||
| BT008873.1 | - | 6 - 7 | 1396 | 61 | ||
More Information
Set Membership: |
|

HD1
Genomic Map