Assay Details
| Gene Symbol: AT4G38340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119996.2 | NP_195547.1 | 1 - 2 | 192 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G38340| Gene Symbol: AT4G38340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119996.2 | NP_195547.1 | 1 - 2 | 192 | 107 | |
Set Membership: |
|