Assay Details
| Gene Symbol: AT4G38340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342491.1 | NP_001329471.1 | 1 - 2 | 789 | 62 | |
| NM_119996.2 | NP_195547.1 | 2 - 3 | 809 | 62 | ||
More Information
Set Membership: |
|

AT4G38340
Genomic Map