Assay Details
| Gene Symbol: PRORP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336377.1 | NP_001324632.1 | 3 - 4 | 1685 | 67 | |
| NM_179855.3 | NP_850186.1 | 3 - 4 | 1463 | 67 | ||
| GenBank mRNA | BT015418.1 | - | 2 - 3 | 1247 | 67 | |
More Information
Set Membership: |
|

PRORP1
Genomic Map