Assay Details
| Gene Symbol: PRORP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336377.1 | NP_001324632.1 | 5 - 6 | 1983 | 142 | |
| NM_179855.3 | NP_850186.1 | 5 - 6 | 1761 | 142 | ||
| GenBank mRNA | BT015418.1 | - | 4 - 5 | 1545 | 142 | |
More Information
Set Membership: |
|

PRORP1
Genomic Map