Assay Details
| Gene Symbol: PHO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179887.3 | NP_850218.1 | 6 - 7 | 3259 | 68 | |
| GenBank mRNA | AY074292.1 | - | 6 - 7 | 3186 | 68 | |
| AY091326.1 | - | 4 - 5 | 2034 | 68 | ||
| DQ027037.1 | - | 4 - 5 | 2034 | 68 | ||
More Information
Set Membership: |
|

PHO2
Genomic Map