Assay Details
| Gene Symbol: AT2G34750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179905.4 | NP_850236.1 | 3 - 4 | 549 | 80 | |
| GenBank mRNA | AY096642.1 | - | 3 - 4 | 411 | 80 | |
| AY142672.1 | - | 2 - 3 | 219 | 80 | ||
More Information
Set Membership: |
|

AT2G34750
Genomic Map