Assay Details
| Gene Symbol: DAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336559.1 | NP_001318358.1 | 1 - 2 | 196 | 66 | |
| NM_129104.4 | NP_565807.1 | 1 - 2 | 192 | 66 | ||
| NM_179916.1 | NP_850247.1 | 1 - 2 | 176 | 66 | ||
| GenBank mRNA | AK119013.1 | - | 1 - 2 | 148 | 66 | |
| AY087959.1 | - | 1 - 2 | 178 | 66 | ||
| BT025589.1 | - | 1 - 2 | 82 | 66 | ||
More Information
Set Membership: |
|

DAD2
Genomic Map