Assay Details
| Gene Symbol: FACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336593.1 | NP_001325233.1 | 5 - 6 | 880 | 79 | |
| NM_179931.4 | NP_850262.2 | 5 - 6 | 880 | 79 | ||
| GenBank mRNA | AJ534971.1 | - | 5 - 6 | 678 | 79 | |
More Information
Set Membership: |
|

FACE2
Genomic Map